missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLYAT Recombinant Rabbit Monoclonal Antibody (4F9J6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558148
This item is not returnable.
View return policy
Description
Immunogen sequence: MMLPLQGAQM LQMLEKSLRK SLPASLKVYG TVFHINHGNP FNLKAVVDKW PDFNTVVVCP QEQDMTDDLD HYTNTYQIYS KDPQNCQEFL GSPELINWKQ.
Mitochondrial acyltransferase which transfers an acyl group to the N-terminus of glycine and glutamine, although much less efficiently. Can conjugate numerous substrates to form a variety of N-acylglycines, with a preference for benzoyl-CoA over phenylacetyl-CoA as acyl donors. Thereby detoxify xenobiotics, such as benzoic acid or salicylic acid, and endogenous organic acids, such as isovaleric acid.
Specifications
| GLYAT | |
| Recombinant Monoclonal | |
| 0.52 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q5PQT3 | |
| GLYAT | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human GLYAT/GAT (NP_964011.2). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 4F9J6 | |
| Unconjugated | |
| GLYAT | |
| A330009E03Rik; AAc; Ab2-132; ACGNAT; Acyl-CoA:glycine N-acyltransferase; AI195249; AI315345; Aralkyl acyl-CoA N-acyltransferase; aralkyl acyl-CoA:amino acid N-acyltransferase; aralkyl-CoA N-acyltransferase; benzoyl-coenzyme A:glycine N-acyltransferase; CAT; GAT; Glyat; Glycine N-acyltransferase; glycine N-benzoyltransferase; glycine-N-acyltransferase; HRP-1(CLP); Liver regeneration-related protein LRRG067 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 293779 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction