missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gly-Phe-Beta-naphthylamide Trifluoroacetic Acid Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Gly-Phe-beta-naphthylamide Trifluoroacetic Acid Salt |
| CAS | 109057-30-9 |
| Molecular Formula | C21H21N3O2 . (C2HF3O2) |
| Quantity | 5 mg |
| Synonym | Glycyl-N-2-naphthalenyl-L-phenylalaninamide Trifluoroacetic Acid Salt, alpha-(2-Aminoacetamido)-N-2-naphthyl-L- hydrocinnamamide Trifluoroacetic Acid Salt, L-Glycylphenyl-N-2-naphthyl Trifluoroacetic Acid Salt, Glycyl-L-phenylalanine-beta-naphthylamide Trifluoroacetic Acid Salt |
| InChI Formula | InChI=1S/C21H21N3O2/c22-14-20(25)24-19(12-15-6-2-1-3-7-15)21(26)23-18-11-10-16-8-4-5-9-17(16)13-18/h1-11,13,19H,12,14,22H2,(H,23,26)(H,24,25)/t19-/m0/s1 |
| SMILES | NCC(=O)N[C@@H](Cc1ccccc1)C(=O)Nc2ccc3ccccc3c2 |
| Molecular Weight (g/mol) | 347.4 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?