missing translation for 'onlineSavingsMsg'
Learn More

Gly-Phe-b-naphthylamide Trifluoroacetic Acid Salt, TRC

Product Code. 30401236
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401236 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401236 Supplier Toronto Research Chemicals Supplier No. TRCG75500525MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gly-Phe-beta-naphthylamide Trifluoroacetic Acid Salt
CAS 109057-30-9
Molecular Formula C21H21N3O2 . (C2HF3O2)
Quantity 25 mg
Synonym Glycyl-N-2-naphthalenyl-L-phenylalaninamide Trifluoroacetic Acid Salt, alpha-(2-Aminoacetamido)-N-2-naphthyl-L- hydrocinnamamide Trifluoroacetic Acid Salt, L-Glycylphenyl-N-2-naphthyl Trifluoroacetic Acid Salt, Glycyl-L-phenylalanine-beta-naphthylamide Trifluoroacetic Acid Salt
InChI Formula InChI=1S/C21H21N3O2/c22-14-20(25)24-19(12-15-6-2-1-3-7-15)21(26)23-18-11-10-16-8-4-5-9-17(16)13-18/h1-11,13,19H,12,14,22H2,(H,23,26)(H,24,25)/t19-/m0/s1
SMILES NCC(=O)N[C@@H](Cc1ccccc1)C(=O)Nc2ccc3ccccc3c2
Molecular Weight (g/mol) 347.4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.