missing translation for 'onlineSavingsMsg'
Learn More

GLXR5 Antibody (4G8), Novus Biologicals™

Product Code. 18323069 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323069 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323069 Supplier Novus Biologicals Supplier No. H00051218M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GLXR5 Monoclonal antibody specifically detects GLXR5 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLXR5
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4G8
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_057501.2
Gene Alias C14orf87chromosome 14 open reading frame 87, FLB4739, glutaredoxin 5, glutaredoxin 5 homolog, glutaredoxin 5 homolog (S. cerevisiae), glutaredoxin-related protein 5, mitochondrial, GRX5, MGC14129, Monothiol glutaredoxin-5, PR01238, PRO1238
Host Species Mouse
Immunogen GLRX5 (NP_057501.2, 1 a.a. ~ 157 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSGSLGRAAAALLRWGRGAGGGGLWGPGVRAAGSGAGGGGSAEQLDALVKKDKVVVFLKGTPEQPQCGFSNAVVQILRLHGVRDYAAYNVLDDPELRQGIKDYSNWPTIPQVYLNGEFVGGCDILLQMHQNGDLVEELKKLGIHSALLDEKKDQDSK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51218
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.