missing translation for 'onlineSavingsMsg'
Learn More

GLX351322, MedChemExpress

Product Code. 16452778
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452778 10 mg 10mg
16445517 5 mg 5mg
16422526 25 mg 25mg
16414236 100 mg 100mg
16409476 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16452778 Supplier Medchem Express Supplier No. HY100111
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GLX351322 is an inhibitor of NADPH oxidase 4 (Nox4), and inhibits hydrogen peroxide production from NOX4-overexpressing cells with an IC50 of 5 μM.

GLX351322 is an inhibitor of NADPH oxidase 4 (Nox4), and inhibits hydrogen peroxide production from NOX4-overexpressing cells with an IC50 of 5 μM.

Specifications

Chemical Name or Material GLX351322
CAS 835598-94-2
Color White
Purity Grade Notes Research
Molecular Formula C21H25N3O5S
Quantity 10 mg
Solubility Information DMSO : 20 mg/mL (46.35 mM; Need ultrasonic)
SMILES O=C(OCC)C1=C(SC2=C1CCC2)NC(CN3CCN(CC3)C(C4=CC=CO4)=O)=O
Molecular Weight (g/mol) 431.51
Formula Weight 431.51
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.