missing translation for 'onlineSavingsMsg'
Learn More

Glutathione, TRC

Product Code. 17983575
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983575 5 g 5g
17941905 1 g 1g
17961465 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17983575 Supplier Toronto Research Chemicals Supplier No. TRCG5979515G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-18-8
Synonym Glutathione∣l-γ-Glutamyl-l-cysteinylglycine

Specifications

Purity >95% (HPLC)
Format Neat
Molecular Weight (g/mol) 307.32
Product Type Metabolite∣API
Quantity 5 g
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.