missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Synthetase Antibody (12F2), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59455C
This item is not returnable.
View return policy
Description
Glutathione Synthetase Monoclonal antibody specifically detects Glutathione Synthetase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| Glutathione Synthetase | |
| Monoclonal | |
| DyLight 650 | |
| EC 6.3.2.3, Glutathione synthase, glutathione synthetase, GSH synthetase, GSHS, GSH-S, MGC14098 | |
| Recombinant human Glutathione Synthetase (1-474aa) purified from E. coli | |
| 0.1 mL | |
| Neuroscience | |
| 2937 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 12F2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction