Learn More
Glutathione S-Transferase pi 1/GSTP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16756
This item is not returnable.
View return policy
Description
Glutathione S-Transferase pi 1/GSTP1 Polyclonal specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Glutathione S-Transferase pi 1/GSTP1 | |
| Western Blot 1:500-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Human, Mouse, Rat | |
| IgG |
| Immunocytochemistry | |
| Affinity Purified | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |