missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione S-transferase Mu 5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97064-50ul
This item is not returnable.
View return policy
Description
Glutathione S-transferase Mu 5 Polyclonal antibody specifically detects Glutathione S-transferase Mu 5 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Glutathione S-transferase Mu 5 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 1-2 μL/mg of lysate, Immunohistochemistry-Paraffin 1:2000-1:10000 | |
| EC 2.5.1.18, glutathione S-alkyltransferase M5, glutathione S-aralkyltransferase M5, glutathione S-aryltransferase M5, glutathione S-transferase M5, glutathione S-transferase mu 5, GST class-mu 5, GSTM5-5, GTM5, S-(hydroxyalkyl)glutathione lyase M5 | |
| Produced in rabbits immunized with purified, recombinant Human Glutathione S-transferase Mu 5 (Accession#: AAH58881.1; Met1-Lys218) | |
| 50 μg | |
| Signal Transduction | |
| 2949 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction