missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione S-transferase Mu 5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35584-20ul
This item is not returnable.
View return policy
Description
Glutathione S-transferase Mu 5 Polyclonal antibody specifically detects Glutathione S-transferase Mu 5 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Glutathione S-transferase Mu 5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 2.5.1.18, glutathione S-alkyltransferase M5, glutathione S-aralkyltransferase M5, glutathione S-aryltransferase M5, glutathione S-transferase M5, glutathione S-transferase mu 5, GST class-mu 5, GSTM5-5, GTM5, S-(hydroxyalkyl)glutathione lyase M5 | |
| A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human Glutathione S-transferase Mu 5 (NP_000842.2).,, Sequence:, LENQVMDNHMELVRLCYDPDFEKLKPKYLEELPEKLKLYSEFLGKRPWFAGDKITFVDFLAYDVLDMKRIFEPKCLDAFLNLKDFISRFEGLKKISAYMKSSQFLRGLLFGKSATWNSK | |
| 20 μL | |
| Signal Transduction | |
| 2949 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction