missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Glutathione S-transferase Mu 5 Antibody (KAB29), Novus Biologicals™
Mouse Monoclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | Glutathione S-transferase Mu 5 |
|---|---|
| Clone | KAB29 |
| Dilution | Western Blot : 1:1000- 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066820
|
Novus Biologicals™
NBP3-30969-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067117
|
Novus Biologicals™
NBP3-30969-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glutathione S-transferase Mu 5 Monoclonal antibody specifically detects Glutathione S-transferase Mu 5 in Broad Species Reactivity samples. It is validated for Western BlotSpecifications
| Glutathione S-transferase Mu 5 | |
| Western Blot : 1:1000- 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Broad Species Reactivity | |
| EC 2.5.1.18, glutathione S-alkyltransferase M5, glutathione S-aralkyltransferase M5, glutathione S-aryltransferase M5, glutathione S-transferase M5, glutathione S-transferase mu 5, GST class-mu 5, GSTM5-5, GTM5, S-(hydroxyalkyl)glutathione lyase M5 | |
| A fusion protein to the internal sequence Glutathione S-transferase Mu 5 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| KAB29 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| Phosphate buffered saline | |
| 2949 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title