missing translation for 'onlineSavingsMsg'
Learn More

Glutathione S-transferase Mu 5 Antibody, DyLight 680 , Novus Biologicals™

Product Code. 18409549 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409549 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409549 Supplier Novus Biologicals Supplier No. NBP297064FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Glutathione S-transferase Mu 5 Polyclonal antibody specifically detects Glutathione S-transferase Mu 5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutathione S-transferase Mu 5
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias EC 2.5.1.18, glutathione S-alkyltransferase M5, glutathione S-aralkyltransferase M5, glutathione S-aryltransferase M5, glutathione S-transferase M5, glutathione S-transferase mu 5, GST class-mu 5, GSTM5-5, GTM5, S-(hydroxyalkyl)glutathione lyase M5
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Glutathione S-transferase Mu 5 (AAH58881.1; Met1-Lys218).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2949
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.