missing translation for 'onlineSavingsMsg'
Learn More

Glutathione S-Transferase mu 1/GSTM1 Antibody [PE], Novus Biologicals™

Product Code. 30733745 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733745 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733745 Supplier Novus Biologicals Supplier No. AF6894P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Glutathione S-Transferase mu 1/GSTM1 Polyclonal antibody specifically detects Glutathione S-Transferase mu 1/GSTM1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutathione S-Transferase mu 1/GSTM1
Applications Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase
Host Species Sheep
Immunogen E. coli-derived recombinant human Glutathione S-Transferase mu 1/GS, Met1-Lys218, Accession # P09488
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2944
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.