missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione S Transferase kappa 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38128-20ul
This item is not returnable.
View return policy
Description
Glutathione S Transferase kappa 1 Polyclonal antibody specifically detects Glutathione S Transferase kappa 1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Glutathione S Transferase kappa 1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| EC 2.5.1.18, glutathione S-transferase k1, glutathione S-transferase kappa 1, Glutathione S-transferase subunit 13, glutathione S-transferase subunit 13 homolog, GST, GST 13-13, GST class-kappa, GST13, GST13-13, GSTK1-1, hGSTK1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-226 of human Glutathione S Transferase kappa 1 (NP_057001.1).,, Sequence:, MGPLPRTVELFYDVLSPYSWLGFEILCRYQNIWNINLQLRPSLITGIMKDSGNKPPGLLPRKGLYMANDLKLLRHHLQIPIHFPKDFLSVMLEKGSLSAMRFLTAVNLEHPEMLEKASRELWMRVWSRNEDITEPQSILAAAEKAGMSAEQAQGLLEKIATPKVKNQLKETTEAACRYGAFGLPITVAHVDGQTHMLFGSDRMELLAHLLGEKWMGPIPPAVNARL | |
| 20 μL | |
| Endocrinology | |
| 373156 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction