missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Reductase Rabbit anti-Human, Clone: S08-2E4, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Glutathione Reductase Monoclonal antibody specifically detects Glutathione Reductase in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Glutathione Reductase |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | S08-2E4 |
| Conjugate | Unconjugated |
| Formulation | 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide |
| Gene Alias | EC 1.8.1, EC 1.8.1.7, GLUR, glutathione reductase, glutathione reductase, mitochondrial, GR, GRase, GRD1, MGC78522 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human Glutathione Reductase (Uniprot#: P00390) |
| Purification Method | Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?