missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 20-187 of Human Glutathione Peroxidase 7 The Recombinant Human Glutathione Peroxidase 7 Protein is derived from E. coli. The Recombinant Human Glutathione Peroxidase 7 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 1 mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. 20 mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 0.1 M NaCl, 1 mM DTT |
| Molecular Weight (g/mol) | 21.8 kDa |
| Purification Method | SDS-PAGE |
| Quantity | 0.5 mg |
| Source | Human |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Common Name | Glutathione Peroxidase 7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?