missing translation for 'onlineSavingsMsg'
Learn More

Glutathione Peroxidase 2/GPX2 Antibody (HL125), Novus Biologicals™

Product Code. 18389895 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389895 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389895 Supplier Novus Biologicals Supplier No. NBP313662

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Glutathione Peroxidase 2/GPX2 Monoclonal antibody specifically detects Glutathione Peroxidase 2/GPX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutathione Peroxidase 2/GPX2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HL125
Conjugate Unconjugated
Formulation PBS
Gene Alias EC 1.11.1, EC 1.11.1.9, Gastrointestinal glutathione peroxidase, gastrointestinal glutathione peroxidase 2, GI-GPx, glutathione peroxidase 2, glutathione peroxidase 2 (gastrointestinal), Glutathione peroxidase-gastrointestinal, Glutathione peroxidase-related protein 2, GPRP, GPRP-2, GPx-2, GPx-GI, GSHPx-2, GSHPx-GI
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Glutathione Peroxidase 2/GPX2. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 2877
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.