missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Disulfide-d10 Ammonium Salt ( >90%), TRC
Specifications
Specifications
| Chemical Name or Material | Glutathione Disulfide-d10 Ammonium Salt |
| Molecular Formula | C20H22D10N6O12S2 • x(NH3) |
| Quantity | 0.5 mg |
| Synonym | N,N'-[Dithiobis[1-[(carboxymethyl)carbamoyl]ethylene]]di-glutamine-d10 Ammonium Salt, L-N,N'-[Dithiobis[1-[(carboxymethyl)carbamoyl]ethylene]]di-glutamine-d10 Ammonium Salt, Glutathione-S-S-glutathione-d10 Ammonium Salt, Glutathione-SSG-d10 Ammonium Salt, Oxidized L-Glutathione-d10 Ammonium Salt, Oxidized Glutathione-d10 Ammonium Salt, Oxiglutatione-d10 Ammonium Salt |
| InChI Formula | InChI=1S/C20H32N6O12S2/c21-9(19(35)36)1-3-13(27)25-11(17(33)23-5-15(29)30)7-39-40-8-12(18(34)24-6-16(31)32)26-14(28)4-2-10(22)20(37)38/h9-12H,1-8,21-22H2,(H,23,33)(H,24,34)(H,25,27)(H,26,28)(H,29,30)(H,31,32)(H,35,36)(H,37,38)/t9-,10?,11-,12-/m0/s1/i1D2,2D2,3D2,4D2,9D,10D |
| SMILES | O=C(O)CNC([C@H](CSSC[C@@H](C(NCC(O)=O)=O)NC(C([2H])([2H])C([2H])([2H])[C@]([2H])(N)C(O)=O)=O)NC(C([2H])([2H])C([2H])([2H])C([2H])(N)C(O)=O)=O)=O |
| Molecular Weight (g/mol) | 622.691703 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?