missing translation for 'onlineSavingsMsg'
Learn More

Glutaryl dichloride, 97%

Product Code. 10102870 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10102870 25 mL Glass bottle 25mL
10676112 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10102870 Supplier Thermo Scientific Acros Supplier No. 119990250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2873-74-7 | C5H6Cl2O2 | 169.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2873-74-7
Molecular Formula C5H6Cl2O2
Molecular Weight (g/mol) 169.01
MDL Number MFCD00000755
InChI Key YVOFTMXWTWHRBH-UHFFFAOYSA-N
Synonym glutaryl dichloride, glutaryl chloride, glutaroyl chloride, glutaric acid dichloride, 1,3-bis chlorocarbonyl propane, glutaroyl dichloride, glutaryl chloride 7ci,8ci, glutaryldichloride, glutarylchloride, glutaric acid chloride
PubChem CID 17887
IUPAC Name pentanedioyl dichloride
SMILES C(CC(=O)Cl)CC(=O)Cl

Specifications

Color Brown-Red or Colorless to Yellow
Density 1.3240g/mL
Boiling Point 216.0°C to 218.0°C
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula ClCO(CH2)3COCl
Refractive Index 1.4720 to 1.4760
Quantity 25 mL
Beilstein 02, 634
Specific Gravity 1.324
Solubility Information Solubility in water: reacts with water.
Formula Weight 169.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Glutaryl dichloride, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Glutaryl dichloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.