missing translation for 'onlineSavingsMsg'
Learn More

Glutaric anhydride, 98%, Thermo Scientific Chemicals

Catalog Number 11449073
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
This item is not returnable. View return policy
108-55-4
C5H6O3
114.1
MFCD00006679
VANNPISTIUFMLH-UHFFFAOYSA-N
glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czech
7940
oxane-2,6-dione
C1CC(=O)OC(=O)C1
This item is not returnable. View return policy
108-55-4
C5H6O3
114.1
MFCD00006679
VANNPISTIUFMLH-UHFFFAOYSA-N
glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czech
7940
oxane-2,6-dione
C1CC(=O)OC(=O)C1

Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.

Solubility
Soluble in Ether, dimethyl sulfoxide and methanol and water.

Notes
Moisture sensitive. Incompatible withstrong oxidizing agents, acids, bases, alcohols and amines.
TRUSTED_SUSTAINABILITY
Melting Point 52°C to 57°C
Density 1.411 g/mL
Boiling Point 291°C
Flash Point 178°C (352°F)
Assay Percent Range 98%
Quantity 50 g
Beilstein 110051
Sensitivity Moisture sensitive
Solubility Information Soluble in Ether,dimethyl sulfoxide and methanol and water.
Formula Weight 114.1
Percent Purity 98%
Chemical Name or Material Glutaric anhydride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'