missing translation for 'onlineSavingsMsg'
Learn More

Glutaric anhydride, 98%

Product Code. 11459073
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459073 250 g 250g
11449073 50 g 50g
11438900 100 g 100g
11439073 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11459073 Supplier Thermo Scientific Alfa Aesar Supplier No. A11152.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-55-4 | C5H6O3 | 114.1 g/mol

Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.

Solubility
Soluble in Ether, dimethyl sulfoxide and methanol and water.

Notes
Moisture sensitive. Incompatible withstrong oxidizing agents, acids, bases, alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-55-4
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.1
MDL Number MFCD00006679
InChI Key VANNPISTIUFMLH-UHFFFAOYSA-N
Synonym glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czech
PubChem CID 7940
IUPAC Name oxane-2,6-dione
SMILES C1CC(=O)OC(=O)C1

Specifications

Melting Point 52°C to 57°C
Density 1.411
Boiling Point 291°C
Flash Point 178°C (352°F)
Assay Percent Range 98%
Quantity 250 g
Beilstein 110051
Sensitivity Moisture sensitive
Solubility Information Soluble in Ether,dimethyl sulfoxide and methanol and water.
Formula Weight 114.1
Percent Purity 98%
Chemical Name or Material Glutaric anhydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.