missing translation for 'onlineSavingsMsg'
Learn More

Glutaric Acid, TRC

Product Code. 17970885
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17970885 5 g 5g
17941685 25 g 25g
17914835 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17970885 Supplier Toronto Research Chemicals Supplier No. TRCG5980005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-94-1
Formula Weight 132.0423
InChI Formula InChI=1S/C5H8O4/c6-4(7)2-1-3-5(8)9/h1-3H2,(H,6,7)(H,8,9)
IUPAC Name pentanedioic acid
Molecular Formula C5 H8 O4
Molecular Weight (g/mol) 132.11
SMILES OC(=O)CCCC(=O)O
Synonym Glutaric Acid, Pentanedioic acid, Glutaric acid (8CI), 1,3-Propanedicarboxylic acid, 1,5-Pentanedioic acid, NSC 9238
Chemical Name or Material Glutaric Acid

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.