missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Glutaredoxin 3/GLRX3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29814
This item is not returnable.
View return policy
Description
Glutaredoxin 3/GLRX3 Polyclonal antibody specifically detects Glutaredoxin 3/GLRX3 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Glutaredoxin 3/GLRX3 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10539 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| FLJ11864, GLRX4, glutaredoxin 3, glutaredoxin 4, glutaredoxin 5, GRX3, GRX4, PICOTbA500G10.4, PKC-interacting cousin of thioredoxin, PKCq-interacting protein, thioredoxin-like 2, TXNL2, TXNL3 | |
| Between 100 and 150 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction