missing translation for 'onlineSavingsMsg'
Learn More

Glutaredoxin 3/GLRX3 Antibody (4B5-2A8), Novus Biologicals™

Product Code. 18320469 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320469 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320469 Supplier Novus Biologicals Supplier No. H00010539M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glutaredoxin 3/GLRX3 Monoclonal antibody specifically detects Glutaredoxin 3/GLRX3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutaredoxin 3/GLRX3
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 4B5-2A8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH05289
Gene Alias FLJ11864, GLRX4, glutaredoxin 3, glutaredoxin 4, glutaredoxin 5, GRX3, GRX4, PICOTbA500G10.4, PKC-interacting cousin of thioredoxin, PKCq-interacting protein, thioredoxin-like 2, TXNL2, TXNL3
Host Species Mouse
Immunogen GLRX3 (AAH05289, 1 a.a. ~ 335 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAGAAEAAVAAVEEVGSAGQFEELLRLKAKSLLVVHFWAPWAPQCAQMNEVMAELAKELPQVSFVKLEAEGVPEVSEKYEISSVPTFLFFKNSQKIDRLDGAHAPELTKKVQRHASSGSFLPSANEHLKEDLNLRLKKLTHAAPCMLFMKGTPQEPRCGFSKQMVEILHKHNIQFSSFDIFSDEEVRQGLKAYSSWPTYPQLYVSGELIGGLDIIKELEASEELDTICPKAPKLEERLKVLTNKASVMLFMKGNKQEAKCGFSKQILEILNSTGVEYETFDILEDEEVRQGLKAYSNWPTYPQLYVKGELVGGLDIVKELKENGELLPILRGEN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10539
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.