missing translation for 'onlineSavingsMsg'
Learn More

Glutaraldehyde, Extra Pure, 50% Solution, SLR, Fisher Chemical

Product Code. 10204630 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10204630 500 mL Plastic Coated Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10204630 Supplier Fisher Chemical Supplier No. G/0518/PB08

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

500mL, Plastic Coated Glass bottle

TRUSTED_SUSTAINABILITY

Specifications

CAS 111-30-8
Melting Point -33°C
pH 3.7
Boiling Point 101.5°C
Packaging Plastic Coated Glass bottle
Molecular Formula C5H8O2
MDL Number MFCD00007025
Quantity 500 mL
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILES O=CCCCC=O
IUPAC Name pentanedial
Molecular Weight (g/mol) 100.12
PubChem CID 3485
Vapor Density 1.05
ChEBI CHEBI:64276
Formula Weight 100.12g/mol
Vapor Pressure 15mmHg at 20°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Glutaraldehyde 50% solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: Glutaraldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.