missing translation for 'onlineSavingsMsg'
Learn More

Glutaraldehyde, 50% aq. soln.

Product Code. 11448743
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11448743

Brand: Thermo Scientific Alfa Aesar A10500.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-30-8 | C5H8O2 | 100.12 g/mol

Glutaraldehyde is used as a cross linking agent for gelatin, poly(vinyl alcohol) and polyheptapeptides. It is also used as a fixative for electron microscopy and as disinfectants. It provides water resistance to protein and polyhydroxy compounds by reacting through cross linking. It acts as an intermediate for the production of resins, dyestuffs, pharmaceuticals, photographic films and for stabilization of proteins on agarose beads, activation of polystyrene and glass for immobilization of antibodies and antigens and coupling peptides onto carrier proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaraldehyde is used as a cross linking agent for gelatin, poly(vinyl alcohol) and polyheptapeptides. It is also used as a fixative for electron microscopy and as disinfectants. It provides water resistance to protein and polyhydroxy compounds by reacting through cross linking. It acts as an intermediate for the production of resins, dyestuffs, pharmaceuticals, photographic films and for stabilization of proteins on agarose beads, activation of polystyrene and glass for immobilization of antibodies and antigens and coupling peptides onto carrier proteins.

Solubility
Miscible with water, acetone, dichloromethane, ethylacetate, isopropanol, n-hexane and toluene.

Notes
Incompatible with strong oxidisng agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-30-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00007025
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
PubChem CID 3485
ChEBI CHEBI:64276
IUPAC Name pentanedial
SMILES O=CCCCC=O

Specifications

Melting Point ∼-21°C
Density 1.123
Boiling Point 100°C
Odor Stench
Linear Formula OHC(CH2)3CHO
Refractive Index 1.421
Quantity 2500 g
UN Number UN2922
Beilstein 605390
Merck Index 14,4472
Solubility Information Miscible with water,acetone,dichloromethane,ethylacetate,isopropanol,n-hexane and toluene.
Formula Weight 100.12
Concentration or Composition (by Analyte or Components) 50% aq. soln.
Chemical Name or Material Glutaraldehyde
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Glutaraldehyde, 50% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Glutaraldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.