missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutaminyl-peptide Cyclotransferase/QPCT Antibody [PerCP], Novus Biologicals™
Shop All Bio Techne ProductsDescription
Glutaminyl-peptide Cyclotransferase/QPCT Polyclonal antibody specifically detects Glutaminyl-peptide Cyclotransferase/QPCT in Human samples. It is validated for Immunohistochemistry
Specifications
Specifications
| Antigen | Glutaminyl-peptide Cyclotransferase/QPCT |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | EC 2.3.2.5, GCT, Glutaminyl cyclase, glutaminyl-peptide cyclotransferase, Glutaminyl-tRNA cyclotransferase, Glutamyl cyclase, QCEC, sQC |
| Host Species | Sheep |
| Immunogen | S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Glutaminyl-peptide Cyclotransferase/QPCT, Ala33-Leu361, Accession # Q16769 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?