missing translation for 'onlineSavingsMsg'
Learn More

Glutaminyl-peptide Cyclotransferase/QPCT Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18318815 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18318815 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18318815 Supplier Novus Biologicals Supplier No. NBP300186AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Glutaminyl-peptide Cyclotransferase/QPCT Polyclonal specifically detects Glutaminyl-peptide Cyclotransferase/QPCT in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutaminyl-peptide Cyclotransferase/QPCT
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 2.3.2.5, GCT, Glutaminyl cyclase, glutaminyl-peptide cyclotransferase, Glutaminyl-tRNA cyclotransferase, Glutamyl cyclase, QCEC, sQC
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Glutaminyl-peptide Cyclotransferase/QPCT (Q16769-1; Ala33-Leu361).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 25797
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.