missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamine synthetase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11037-2-AP-20UL
This item is not returnable.
View return policy
Description
Structural maintece of chromosomes (SMC) and non-SMC condensin proteins associate into complexes that have been implicated in the process of chromosome condensation. A crucial prerequisite for accurate segregation of replicated sister chromatids is the condensation of the chromosomes into a manageable form prior to metaphase. The condensin I complex consists of two SMC subunits, SMC2 and SMC4, and three non-SMC subunits, CAP-H, CAP-G, and CAP-D2. An alternative complex, the condensin II complex, contains alternate non-SMC subunits, CAP-G2, CAP-H2, and CAP-D3. CAP-H2 is also known as Non-SMC condensin II complex, subunit H2 (NCAPH2) or kleisin beta isoform 2. The three non-SMC subunits in the condensing complexes form a regulatory subcomplex that is required to activate the SMC ATPases and to promote mitosis-specific chromatin binding of the holocomplex. The precise individual functions of each non-SMC protein in activation remain to be determined.Specifications
| Glutamine synthetase | |
| Polyclonal | |
| Unconjugated | |
| GLUL | |
| GLNS, GLUL, Glutamate ammonia ligase, Glutamate decarboxylase, Glutamine synthetase, GS, PIG43, PIG59 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14645, 24957, 2752 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry, Flow Cytometry | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P09606, P15104, P15105 | |
| GLUL | |
| Glutamine synthetase Fusion Protein Ag1510 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction