missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamine Synthetase Polyclonal Antibody
Glutamine Synthetase Polyclonal Antibody
Brand: Invitrogen PA581505
This item is not returnable.
View return policy
Description
Glutamine Synthetase Polyclonal Antibody for Western Blot, IHC (P), IP
Specifications
| Glutamine Synthetase | |
| Polyclonal | |
| Unconjugated | |
| GLUL | |
| cell proliferation-inducing protein 59; GLNA; GLNS; Glul; GLUL protein; Glutamate ammonia ligase; glutamate decarboxylase; glutamate-ammonia ligase; glutamate--ammonia ligase; glutamate-ammonia ligase (glutamine synthase); glutamate-ammonia ligase (glutamine synthetase); glutamine synthase; glutamine synthetase; Glutamine synthetase (glutamate-ammonia ligase); glutamine synthetase 1; glutamine synthetase I; GS; Palmitoyltransferase GLUL; PIG43; PIG59; Proliferation inducing protein 43; proliferation-inducing protein 43 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 2752 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| P15104 | |
| GLUL | |
| A synthetic peptide corresponding to the C-terminus of the Human GLUL. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction