missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamine synthetase Mouse anti-Human, Porcine, Clone: 1D10G8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66323-1-IG-20UL
This item is not returnable.
View return policy
Description
Structural maintece of chromosomes (SMC) and non-SMC condensin proteins associate into complexes that have been implicated in the process of chromosome condensation. A crucial prerequisite for accurate segregation of replicated sister chromatids is the condensation of the chromosomes into a manageable form prior to metaphase. The condensin I complex consists of two SMC subunits, SMC2 and SMC4, and three non-SMC subunits, CAP-H, CAP-G, and CAP-D2. An alternative complex, the condensin II complex, contains alternate non-SMC subunits, CAP-G2, CAP-H2, and CAP-D3. CAP-H2 is also known as Non-SMC condensin II complex, subunit H2 (NCAPH2) or kleisin beta isoform 2. The three non-SMC subunits in the condensing complexes form a regulatory subcomplex that is required to activate the SMC ATPases and to promote mitosis-specific chromatin binding of the holocomplex. The precise individual functions of each non-SMC protein in activation remain to be determined.Specifications
| Glutamine synthetase | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P15104, P46410 | |
| GLUL | |
| Glutamine synthetase Fusion Protein Ag6309 | |
| 20 μL | |
| Primary | |
| Pig, Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1D10G8 | |
| Unconjugated | |
| GLUL | |
| GLNS, GLUL, Glutamate ammonia ligase, Glutamate decarboxylase, Glutamine synthetase, GS, PIG43, PIG59 | |
| Mouse | |
| Protein A | |
| RUO | |
| 2752, 396944 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction