missing translation for 'onlineSavingsMsg'
Learn More

Glutamine Synthetase Mouse anti-Human, Bovine, Equine, Clone: GLUL/6600, Novus Biologicals™

Product Code. 18340475 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340475 20 μg 20µg
18370494 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18340475 Supplier Novus Biologicals Supplier No. NBP31392820UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Human, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamine Synthetase
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GLUL/6600
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P15104
Gene Alias cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43
Host Species Mouse
Immunogen Recombinant fragment (around aa 50-250) of human Glutamine Synthetase protein (exact sequence is proprietary) (Uniprot: P15104)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Hypoxia, Neurodegeneration, Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2752
Target Species Human, Bovine, Equine
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2bc κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.