missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamine Synthetase Antibody (GLUL/6600), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14023H
This item is not returnable.
View return policy
Description
Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Human, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Glutamine Synthetase | |
| Monoclonal | |
| HRP | |
| cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43 | |
| Recombinant fragment (around aa 50-250) of human Glutamine Synthetase protein (exact sequence is proprietary) (Uniprot: P15104) | |
| 0.1 mL | |
| Hypoxia, Neurodegeneration, Neuroscience, Vision | |
| 2752 | |
| Store at 4°C in the dark. | |
| IgG2bc κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| GLUL/6600 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Bovine, Equine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction