missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamine Synthetase Antibody (CL3004) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46644G
This item is not returnable.
View return policy
Description
Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
Specifications
| Glutamine Synthetase | |
| Monoclonal | |
| DyLight 488 | |
| cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43 | |
| This antibody was developed using a recombinant protein derived from P15104, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Hypoxia, Neurodegeneration, Neuroscience, Vision | |
| 2752 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CL3004 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction