missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamine (L-Glutamine), LoGiCal
Description
Glutamine (L-Glutamine)
Specifications
Specifications
| Chemical Name or Material | L-Glutamine |
| CAS | 56-85-9 |
| Analyte or Component Names | L-Glutamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H10 N2 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-Glutamine, (2 S)-2,5-Diamino-5-oxopentanoic Acid, Aspartic Acid Imp. F (Pharmeuropa), 2-Aminoglutaramic acid, Acustasin, Aesgen 14, Cebrogen, Glumin, Glumin (amino acid), Glutamic acid 5-amide, Glutamic acid amide, Glutamine, L-(+)-Glutamine, L-2-Aminoglutaramidic acid, Levoglutamide, NSC 27421, (S)-2,5-Diamino-5-oxopentanoic acid, Saforis, Stimulina, L-Glutamic acid gamma-amide, gamma-Glutamine, Asparagine Monohydrate Impurity E, Aspartic Acid Impurity F, Magnesium Aspartate Dihydrate Impurity G, Asparagine Impurity E, Magnesium Aspartate Impurity G |
| InChI Formula | InChI=1 S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1 |
| SMILES | N[C@@H](CCC(=O)N)C(=O)O |
| IUPAC Name | (2 S)-2,5-diamino-5-oxopentanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?