missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Glutaminase Monoclonal antibody specifically detects Glutaminase in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
Specifications
| Antigen | Glutaminase |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Clone | GT1075 |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | AAD20, EC 3.5.1.2, GLS1DKFZp686O15119, glutaminase, glutaminase C, glutaminase kidney isoform, mitochondrial, glutaminase, phosphate-activated, K-glutaminase, KIAA0838FLJ10358, L-glutamine amidohydrolase |
| Host Species | Mouse |
| Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Glutaminase (GAC). The exact sequence is proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?