missing translation for 'onlineSavingsMsg'
Learn More

Glutaminase Antibody (5C4), Novus Biologicals™

Product Code. 18368098 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368098 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368098 Supplier Novus Biologicals Supplier No. H00002744M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glutaminase Monoclonal antibody specifically detects Glutaminase in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutaminase
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 5C4
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055720
Gene Alias AAD20, EC 3.5.1.2, GLS1DKFZp686O15119, glutaminase, glutaminase C, glutaminase kidney isoform, mitochondrial, glutaminase, phosphate-activated, K-glutaminase, KIAA0838FLJ10358, L-glutamine amidohydrolase
Host Species Mouse
Immunogen GLS (NP_055720, 580 a.a. ~ 669 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EQRDYDSRTALHVAAAEGHVEVVKFLLEACKVNPFPKDRWNNTPMDEALHFGHHDVFKILQEYQVQYTPQGDSDNGKENQTVHKNLDGLL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2744
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.