missing translation for 'onlineSavingsMsg'
Learn More

Glutamic Acid (L-Glutamic Acid), Mikromol™

Product Code. 17949644
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17949644 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17949644 Supplier Mikromol Supplier No. MM1333.00
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Glutamic Acid (L-Glutamic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Glutamic acid
CAS 56-86-0
Analyte or Component Names L-Glutamic acid
Shipping Condition Room Temperature
Molecular Formula C5 H9 N O4
Certifications/Compliance ISO 17034
Synonym Glutamic acid, L-Glutamic acid, Glutamic acid, L- (7CI,8CI), (+)-L-Glutamic acid, (2S)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-2-Amino-1,5-pentanedioic acid, (S)-2-Aminopentanedioic acid, (S)-Glutamic acid, 1-Aminopropane-1,3-dicarboxylic acid, 2-Aminoglutaric acid, 2-Aminopentanedioic acid, Aciglut, E 620, Glusate, Glutacid, Glutamicol, Glutamidex, Glutaminic acid, Glutaminol, Glutaton, L-(+)-Glutamic acid, L-Glutaminic acid, NSC 143503, Pentanedioic acid, 2-amino-, (S)-, Sortem, l-Glutaminic acid, L-alpha-Aminoglutaric acid, alpha-Aminoglutaric acid, alpha-Glutamic acid, Alanine Impurity B, Asparagine Monohydrate Impurity B, Asparagine Monohydrate Impurity B, Aspartic Acid Impurity C, Magnesium Aspartate Dihydrate Impurity D, Magnesium Pidolate Impurity A, Asparagine Impurity B, Magnesium Aspartate Impurity D
InChI Formula InChI=1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1
SMILES N[C@@H](CCC(=O)O)C(=O)O
IUPAC Name (2S)-2-aminopentanedioic acid
Molecular Weight (g/mol) 147.13
Quantity 250 mg
Formula Weight 147.0532 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.