missing translation for 'onlineSavingsMsg'
Learn More

Glutamic Acid (L-Glutamic Acid), LoGiCal

Product Code. 30458936
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458936 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458936 Supplier LoGiCal Supplier No. LGCFOR1333.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Glutamic Acid (L-Glutamic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Glutamic acid
CAS 56-86-0
Analyte or Component Names L-Glutamic acid
Shipping Condition Room Temperature
Molecular Formula C5 H9 N O4
Certifications/Compliance ISO 9001
Synonym Glutamic acid, L-Glutamic acid, Glutamic acid, L- (7 CI,8 CI), (+)-L-Glutamic acid, (2 S)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-2-Amino-1,5-pentanedioic acid, (S)-2-Aminopentanedioic acid, (S)-Glutamic acid, 1-Aminopropane-1,3-dicarboxylic acid, 2-Aminoglutaric acid, 2-Aminopentanedioic acid, Aciglut, E 620, Glusate, Glutacid, Glutamicol, Glutamidex, Glutaminic acid, Glutaminol, Glutaton, L-(+)-Glutamic acid, L-Glutaminic acid, NSC 143503, Pentanedioic acid, 2-amino-, (S)-, Sortem, l-Glutaminic acid, L-alpha-Aminoglutaric acid, alpha-Aminoglutaric acid, alpha-Glutamic acid, Alanine Impurity B, Asparagine Monohydrate Impurity B, Asparagine Monohydrate Impurity B, Aspartic Acid Impurity C, Magnesium Aspartate Dihydrate Impurity D, Magnesium Pidolate Impurity A, Asparagine Impurity B, Magnesium Aspartate Impurity D
InChI Formula InChI=1 S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1
SMILES N[C@@H](CCC(=O)O)C(=O)O
IUPAC Name (2 S)-2-aminopentanedioic acid
Industry Type Forensics
Molecular Weight (g/mol) 147.13
Quantity 10 mg
Formula Weight 147.0532 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.