missing translation for 'onlineSavingsMsg'
Learn More

Glutamate Receptor 4 Rabbit anti-Mouse, Rat, Clone: 4M5D0, Novus Biologicals™

Product Code. 18319631 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319631 100 μg 100µL
18354157 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18319631 Supplier Novus Biologicals Supplier No. NBP316445100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Glutamate Receptor 4 Monoclonal antibody specifically detects Glutamate Receptor 4 in Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamate Receptor 4
Applications Western Blot
Classification Monoclonal
Clone 4M5D0
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias AMPA-selective glutamate receptor 4, GluA4, GluR4, GluR-4, GLUR4C, GLUR4gluR-D, GluR-D, GLURD, glutamate receptor 4, Glutamate receptor ionotropic, AMPA 4, glutamate receptor, ionotrophic, AMPA 4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 800-902 of human Glutamate Receptor 4 (P48058).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Alzheimers Research, GPCR, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2893
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.