missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamate Receptor 4 Antibody (1059705), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB113631PECY55
This item is not returnable.
View return policy
Description
Glutamate Receptor 4 Monoclonal antibody specifically detects Glutamate Receptor 4 in Human samples. It is validated for Flow Cytometry
Specifications
| Glutamate Receptor 4 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1059705 | |
| Flow Cytometry | |
| AMPA-selective glutamate receptor 4, GluA4, GluR4, GluR-4, GLUR4C, GLUR4gluR-D, GluR-D, GLURD, glutamate receptor 4, Glutamate receptor ionotropic, AMPA 4, glutamate receptor, ionotrophic, AMPA 4 | |
| Synthetic peptide, Accession # P48058 | |
| 0.1 mL | |
| Alzheimers Research, GPCR, Neurodegeneration, Neuroscience, Signal Transduction | |
| 2893 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction