missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamate Dehydrogenase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB600-853
This item is not returnable.
View return policy
Description
Glutamate Dehydrogenase Polyclonal specifically detects Glutamate Dehydrogenase in Human, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation.
Specifications
| Glutamate Dehydrogenase | |
| Polyclonal | |
| Western Blot 1:1000-1:3000, ELISA 1:4000-1:16000, Immunohistochemistry, Immunoprecipitation 1:100 | |
| P00366 | |
| GLUD1 | |
| This Glutamate Dehydrogenase Antibody was prepared from whole rabbit serum produced by repeated immunizations with a full length Glutamate Dehydrogenase protein isolated from Bovine Liver. (Uniprot: P00366) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross-reacts with Cow. Not yet tested in other species. | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| EC 1.4.1, EC 1.4.1.3, GDH, GDH 1, GDH1, GLUD, glutamate dehydrogenase (NAD(P)+), glutamate dehydrogenase 1, glutamate dehydrogenase 1, mitochondrial, MGC132003 | |
| Rabbit | |
| 61.51 kDa | |
| 0.2 mL | |
| Cellular Markers, Mitochondrial Markers, Neuroscience | |
| 2746 | |
| Human, Bovine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction