Learn More
Glutamate Dehydrogenase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-54700
 View more versions of this product
Product Code. 18221740
  • £488.00 / 100µL
Description
Glutamate Dehydrogenase Polyclonal antibody specifically detects Glutamate Dehydrogenase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glutamate Dehydrogenase
Polyclonal
Immunohistochemistry-Paraffin 1:500 - 1:1000
EC 1.4.1, EC 1.4.1.3, GDH, GDH 1, GDH1, GLUD, glutamate dehydrogenase (NAD(P)+), glutamate dehydrogenase 1, glutamate dehydrogenase 1, mitochondrial, MGC132003
Rabbit
Affinity Purified
RUO
Primary
Specificity of human Glutamate Dehydrogenase antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunohistochemistry (Paraffin)
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
GLUD1
This antibody was developed against a Recombinant Protein corresponding to amino acids:KPCNHVLSLSFPIRRDDGSWEVIEGYRAQHSQHRTPCKGGIRYSTDVSVDEVKALASLMTYKCAVVDVPFGGAKAGVKINPKN
100 μL
Cellular Markers, Diabetes Research, Mitochondrial Markers, Neuroscience
2746