missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamate Dehydrogenase 2/GLUD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38295-100ul
This item is not returnable.
View return policy
Description
Glutamate Dehydrogenase 2/GLUD2 Polyclonal antibody specifically detects Glutamate Dehydrogenase 2/GLUD2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Glutamate Dehydrogenase 2/GLUD2 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| EC 1.4.1, EC 1.4.1.3, GDH 2, GDH2, GLUDP1, glutamate dehydrogenase 2, glutamate dehydrogenase 2, mitochondrial, glutamate dehydrogenase pseudogene 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 319-558 of human Glutamate Dehydrogenase 2/GLUD2 (NP_036216.2).,, Sequence:, YLHRFGAKCIAVGESDGSIWNPDGIDPKELEDFKLQHGSILGFPKAKPYEGSILEVDCDILIPAATEKQLTKSNAPRVKAKIIAEGANGPTTPEADKIFLERNILVIPDLYLNAGGVTVSYFEWLKNLNHVSYGRLTFKYERDSNYHLLLSVQESLERKFGKHGGTIPIVPTAEFQDSISGASEKDIVHSALAYTMERSARQIMHTAMKYNLGLDLRTAAYVNAIEKVFKVYSEAGVTFT | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2747 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction