missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLUT9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522960
This item is not returnable.
View return policy
Description
Suggested positive control: human kidney membrane protein.
GLUT9, or SLC2A9, is a member of the SLC2A facilitative glucose transporter family. Members of this family play a significant role in maintaining glucose homeostasis. GLUT9 may play a role in the development and survival of chondrocytes in cartilage matrices. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| GLUT9 | |
| Polyclonal | |
| Unconjugated | |
| SLC2A9 | |
| facilitated glucose transporter, type 9; glucose transporter type 9; GLUT9; GLUT-9; GLUTX; human glucose transporter-like protein-9; Slc2a9; SLC2a9A; SLC2A9B; solute carrier family 2 (facilitated glucose transporter), member 9; solute carrier family 2 member 9; solute carrier family 2, facilitated glucose transporter member 9; UAQTL2; Urate transporter; urate voltage-driven efflux transporter 1; URATv1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117591, 56606 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q3T9X0, Q9NRM0 | |
| SLC2A9 | |
| Synthetic peptide made to an internal portion of human GLUT9 (within residues 500-550). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction