missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLUT9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564269
This item is not returnable.
View return policy
Description
Immunogen sequence: NRNSKELGLV PLTDDTSHAR PPGPGRALLE CDHLRSGVPG GRRRKDWS Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 31%.
GLUT9, or SLC2A9, is a member of the SLC2A facilitative glucose transporter family. Members of this family play a significant role in maintaining glucose homeostasis. GLUT9 may play a role in the development and survival of chondrocytes in cartilage matrices. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| GLUT9 | |
| Polyclonal | |
| Unconjugated | |
| SLC2A9 | |
| facilitated glucose transporter, type 9; glucose transporter type 9; GLUT9; GLUT-9; GLUTX; human glucose transporter-like protein-9; Slc2a9; SLC2a9A; SLC2A9B; solute carrier family 2 (facilitated glucose transporter), member 9; solute carrier family 2 member 9; solute carrier family 2, facilitated glucose transporter member 9; UAQTL2; Urate transporter; urate voltage-driven efflux transporter 1; URATv1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56606 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NRM0 | |
| SLC2A9 | |
| Recombinant protein corresponding to Human GLUT9. Recombinant protein control fragment (Product #RP-103626). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction