missing translation for 'onlineSavingsMsg'
Learn More

GLUT9 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18421814 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421814 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421814 Supplier Novus Biologicals Supplier No. NBP106565FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GLUT9 Polyclonal antibody specifically detects GLUT9 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT9
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias facilitated glucose transporter member 9, GLUT-9, GLUT9Glut9, GLUTX, solute carrier family 2 (facilitated glucose transporter), member 9, UAQTL2, urate voltage-driven efflux transporter 1
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human GLUT9 (within residues 10-60). [Swiss-Prot# Q9NRM0]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 56606
Target Species Human, Mouse, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.