missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLUT9 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-06565R
This item is not returnable.
View return policy
Description
GLUT9 Polyclonal specifically detects GLUT9 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GLUT9 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry-Paraffin | |
| Q9NRM0 | |
| SLC2A9 | |
| Synthetic peptide made to an internal portion of human GLUT9 (within residues 10-60). [Swiss-Prot# Q9NRM0] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for the human GLUT9 protein Isoform 1 (SLC2A9-L). | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| facilitated glucose transporter member 9, GLUT-9, GLUT9Glut9, GLUTX, solute carrier family 2 (facilitated glucose transporter), member 9, UAQTL2, urate voltage-driven efflux transporter 1 | |
| Rabbit | |
| 58.7 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 56606 | |
| Human, Mouse, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction