missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLUT5 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142485
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
Cytochalasin B-sensitive carrier. Seems to function primarily as a fructose transporter.
Specifications
| GLUT5 | |
| Polyclonal | |
| Unconjugated | |
| SLC2A5 | |
| AI526984; Fructose transporter; glucose transporter type 5, small intestine; glucose transporter-like protein 5; GLUT5; GLUT-5; RP5-963K15.1; SLC2A5; Slc5a; solute carrier family 2 (facilitated glucose transporter), member 5; solute carrier family 2 (facilitated glucose/fructose transporter), member 5; solute carrier family 2 member 5; solute carrier family 2, facilitated glucose transporter member 5; solute carrier family 2, member 5; testicular tissue protein Li 81 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 6518 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P22732 | |
| SLC2A5 | |
| Peptide with sequence C-SEVYPEKEELKE. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction