missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antibody detects endogenous levels of total Glucose Transporter 5 GLUT5.
Cytochalasin B-sensitive carrier. Seems to function primarily as a fructose transporter.
Specifications
Specifications
| Antigen | GLUT5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | SLC2A5 |
| Gene Accession No. | P22732, P43427, Q9WV38 |
| Gene Alias | AI526984; Fructose transporter; glucose transporter type 5, small intestine; glucose transporter-like protein 5; GLUT5; GLUT-5; RP5-963K15.1; SLC2A5; Slc5a; solute carrier family 2 (facilitated glucose transporter), member 5; solute carrier family 2 (facilitated glucose/fructose transporter), member 5; solute carrier family 2 member 5; solute carrier family 2, facilitated glucose transporter member 5; solute carrier family 2, member 5; testicular tissue protein Li 81 |
| Gene Symbols | SLC2A5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
